Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138356.2 | NP_612365.2 | 4, 5 | 1143 | ||
XM_006720742.3 | XP_006720805.2 | 3, 4 | 858 | |||
XM_011522174.2 | XP_011520476.1 | 3, 4 | 858 | |||
XM_011522175.2 | XP_011520477.1 | 3, 4 | 858 | |||
XM_017022710.1 | XP_016878199.1 | 3, 4 | 859 | |||
XM_017022711.1 | XP_016878200.1 | 3, 4 | 969 | |||
GenBank mRNA | AK094391.1 | - | 3, 4 | 587 | ||
AK096267.1 | - | 4, 5 | 1143 | |||
BC101601.1 | - | 2, 3 | 299 | 8069107 | ||
BC101603.1 | - | 2, 3 | 298 | 8069109 | ||
BC143601.1 | - | 2, 3 | 299 |