Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198963.1 | NP_945314.1 | 5 | 1357 | ||
XM_011533154.2 | XP_011531456.1 | 5 | 1361 | |||
XM_011533155.2 | XP_011531457.1 | 5 | 1359 | |||
XM_011533156.2 | XP_011531458.1 | 5 | 1361 | |||
XM_011533157.1 | XP_011531459.1 | 5 | 1361 | |||
GenBank mRNA | AF283512.1 | - | 1 | 48 | ||
AK057423.1 | - | 4 | 1157 | |||
AK296739.1 | - | 5 | 1396 | |||
AK307514.1 | - | 5 | 1386 | |||
BC039270.1 | - | 4 | 1164 | |||
BC060778.1 | - | 5 | 1357 | |||
BC131534.1 | - | 5 | 1329 | 40082400 | ||
BQ719095.1 | - | 1 | 220 |