Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SESTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178123.4 | NP_835224.3 | 18 | 2175 | ||
XM_011512139.2 | XP_011510441.1 | 18 | 2203 | |||
XM_011512141.2 | XP_011510443.1 | 16 | 1880 | |||
XM_017005250.1 | XP_016860739.1 | 17 | 2029 | |||
XM_017005251.1 | XP_016860740.1 | 19 | 2381 | |||
GenBank mRNA | AK057944.1 | - | 6 | 644 | ||
AK096232.1 | - | 18 | 2153 | |||
AL122046.1 | - | 8 | 1056 | |||
AY209190.1 | - | 18 | 2072 | |||
BC029744.1 | - | 5 | 578 | |||
BC047578.1 | - | 18 | 2324 | 4815542 | ||
BC053862.1 | - | 6 | 627 |