Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACBD5 SIRNA Assays ›
Gene Symbol:
ACBD5
Gene Name:
acyl-CoA binding domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.10: 27182624 - 27248674 on Build GRCh38
UniGene ID:
Hs.530597
Species:
Human
siRNA ID
HSS132158
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ACBD5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042473.3
NP_001035938.1
9
1238
NM_001271512.3
NP_001258441.1
8
1552
NM_001301251.1
NP_001288180.1
8
1117
NM_001301252.1
NP_001288181.1
7
1431
NM_001301253.1
NP_001288182.1
8
1107
NM_001301254.1
NP_001288183.1
6
1227
NM_145698.4
NP_663736.2
9
1363
XM_006717528.3
XP_006717591.3
10
7796
XM_006717530.3
XP_006717593.1
10
1395
XM_006717531.2
XP_006717594.1
10
1405
XM_006717535.2
XP_006717598.1
10
1262
XM_017016884.1
XP_016872373.1
10
7796
XM_017016885.1
XP_016872374.1
9
7763
XM_017016886.1
XP_016872375.1
9
7763
XM_017016887.1
XP_016872376.1
10
1326
XM_017016888.1
XP_016872377.1
10
1391
XM_017016889.1
XP_016872378.1
9
1293
XM_017016890.1
XP_016872379.1
9
7592
XM_017016891.1
XP_016872380.1
9
1358
XM_017016892.1
XP_016872381.1
9
1279
XM_017016893.1
XP_016872382.1
8
7559
XM_017016894.1
XP_016872383.1
8
7559
XM_017016895.1
XP_016872384.1
10
1262
XM_017016896.1
XP_016872385.1
9
1230
XM_017016897.1
XP_016872386.1
9
1187
XM_017016898.1
XP_016872387.1
8
1154
XM_017016899.1
XP_016872388.1
8
1154
XM_017016900.1
XP_016872389.1
9
1180
XM_017016901.1
XP_016872390.1
9
1160
XM_017016902.1
XP_016872391.1
9
1179
XM_017016903.1
XP_016872392.1
9
1164
XM_017016904.1
XP_016872393.1
8
1159
XM_017016905.1
XP_016872394.1
8
1145
XM_017016906.1
XP_016872395.1
8
1026
XM_017016907.1
XP_016872396.1
8
1159
XM_017016908.1
XP_016872397.1
8
1148
GenBank mRNA
AK057469.1
-
9
1230
AK294510.1
-
6
1145
AK295031.1
-
8
1189
AL133064.1
-
3
446
BC030555.1
-
9
1267
5218685
Back To Top
More Information
Panther Classification:
Molecular Function -
transfer/carrier protein
Gene Ontology Categories:
Function(s)
Process(es)
transport
macroautophagy
pexophagy
aggrephagy
fatty-acyl-CoA binding
molecular_function
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline