Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080539.1 | NP_001074008.1 | 19, 20 | 2296 | ||
XM_006712438.3 | XP_006712501.1 | 18, 19 | 2233 | |||
XM_006712439.3 | XP_006712502.1 | 3, 4 | 880 | |||
XM_011510985.2 | XP_011509287.1 | 17, 18 | 2025 | |||
XM_011510987.2 | XP_011509289.1 | 11, 12 | 1216 | |||
XM_011510988.2 | XP_011509290.1 | 15, 16 | 1742 | |||
XM_017003861.1 | XP_016859350.1 | 17, 18 | 2140 | |||
XM_017003862.1 | XP_016859351.1 | 17, 18 | 2119 | |||
XM_017003863.1 | XP_016859352.1 | 18, 19 | 2233 | |||
XM_017003864.1 | XP_016859353.1 | 18, 19 | 2233 | |||
XM_017003865.1 | XP_016859354.1 | 15, 16 | 1757 | |||
XM_017003866.1 | XP_016859355.1 | 12, 13 | 1432 | |||
GenBank mRNA | AK096130.1 | - | 1, 2 | 384 | ||
AK301665.1 | - | 4, 5 | 484 | |||
AK302685.1 | - | 14, 15 | 1664 | |||
AL834537.1 | - | 3, 4 | 775 | |||
BC062449.1 | - | 1, 2 | 376 | |||
BC062658.1 | - | 3, 4 | 836 |