Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMSAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043318.2 | NP_001036783.1 | 8 | 2865 | ||
NM_194278.3 | NP_919254.2 | 8 | 3244 | |||
XM_005268204.1 | XP_005268261.1 | 8 | 3244 | |||
XM_005268205.1 | XP_005268262.1 | 8 | 2800 | |||
XM_005268206.1 | XP_005268263.1 | 9 | 3017 | |||
GenBank mRNA | AK090424.1 | - | 3 | 2502 | ||
AK127785.1 | - | 9 | 3243 | |||
BC003121.1 | - | 8 | 2037 | |||
BC006451.1 | - | 8 | 1699 | |||
BC006511.2 | - | 8 | 2495 | |||
BC009202.2 | - | 9 | 2865 | |||
BC015668.2 | - | 8 | 2878 | |||
BC025330.1 | - | 8 | 2495 |