Gene Symbol: TIMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001563.2 | NP_001001563.1 | 5 | 782 | ||
XM_011527491.2 | XP_011525793.1 | 5 | 325 | |||
GenBank mRNA | AY551341.1 | - | 5 | 340 | ||
BC009072.1 | - | 5 | 314 | |||
BC010736.1 | - | 5 | 335 | |||
BC050082.1 | - | 5 | 358 | |||
BC121147.1 | - | 5 | 710 | 40121258 | ||
BQ925812.1 | - | 5 | 369 |