Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139072.3 | NP_620711.3 | 2 | 704 | ||
XM_005246950.2 | XP_005247007.1 | 2 | 691 | |||
GenBank mRNA | AF442487.1 | - | 2 | 617 | ||
AK289553.1 | - | 2 | 662 | |||
AY358891.1 | - | 2 | 667 | |||
BC035009.1 | - | 2 | 694 | 4820343 | ||
DA400901.1 | - | 2 | 444 |