Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145236.2 | NP_660279.1 | 2 | 1221 | ||
XM_005246953.1 | XP_005247010.1 | 2 | 1130 | |||
GenBank mRNA | AF502430.1 | - | 2 | 1221 | ||
AK000770.1 | - | 1 | 405 | |||
AK126207.1 | - | 2 | 1117 |