Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA2D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172364.4 | NP_758952.4 | 13 | 1635 | ||
XM_011521041.2 | XP_011519343.1 | 13 | 5546 | |||
GenBank mRNA | AF516695.1 | - | 13 | 1767 | ||
BC150186.1 | - | 13 | 1568 | 40148321 | ||
BX537436.1 | - | 13 | 1598 | |||
BX537437.1 | - | 13 | 1636 |