Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297559.1 | NP_001284488.1 | 3 | 731 | ||
NM_053044.4 | NP_444272.1 | 3 | 731 | |||
XM_011513596.2 | XP_011511898.1 | 3 | 739 | |||
GenBank mRNA | AL547377.3 | - | 3 | 734 | ||
AY040094.2 | - | 3 | 748 | |||
AY280665.1 | - | 3 | 719 | |||
AY280666.1 | - | 3 | 634 | |||
BC034390.1 | - | 3 | 731 | 5180585 | ||
BC035717.1 | - | 3 | 731 | 5588608 |