Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033226.2 | NP_150229.2 | 5 | 1081 | ||
GenBank mRNA | AF395908.1 | - | 5 | 741 | ||
AF395909.1 | - | 5 | 741 | |||
AF411577.1 | - | 5 | 741 | |||
AF411578.1 | - | 5 | 741 | |||
AK122635.1 | - | 6 | 938 | |||
AK302520.1 | - | 6 | 938 | |||
AY040220.1 | - | 5 | 1081 | |||
BC036378.1 | - | 7 | 1239 |