Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138409.2 | NP_612418.2 | 4 | 734 | ||
XM_011535400.2 | XP_011533702.1 | 4 | 660 | |||
XM_017010219.1 | XP_016865708.1 | 5 | 1032 | |||
XM_017010220.1 | XP_016865709.1 | 4 | 1301 | |||
GenBank mRNA | AK090775.1 | - | 4 | 734 | ||
AK292736.1 | - | 4 | 665 | |||
AK307078.1 | - | 3 | 563 | |||
BC010003.2 | - | 4 | 679 | 4134852 | ||
BC039855.1 | - | 4 | 597 | 3850270 |