Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102564.1 | NP_001096034.1 | 2 | 144 | ||
NM_001255995.1 | NP_001242924.1 | 2 | 144 | |||
NM_052873.2 | NP_443105.2 | 2 | 144 | |||
NR_045664.1 | 2 | 144 | ||||
NR_045665.1 | 2 | 144 | ||||
GenBank mRNA | AK056735.1 | - | 2 | 117 | ||
AK302944.1 | - | 2 | 127 | |||
BC010436.1 | - | 2 | 115 | 3608220 | ||
BI821833.1 | - | 2 | 120 | |||
CB853974.1 | - | 2 | 105 | |||
DB443802.1 | - | 2 | 144 | |||
DB497730.1 | - | 2 | 127 |