Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282761.1 | NP_001269690.1 | 4 | 529 | ||
NM_080667.6 | NP_542398.3 | 3 | 454 | |||
XM_017003254.1 | XP_016858743.1 | 3 | 445 | |||
XM_017003255.1 | XP_016858744.1 | 3 | 468 | |||
GenBank mRNA | AK308776.1 | - | 2 | 310 | ||
AK309764.1 | - | 3 | 403 | |||
AY358097.1 | - | 4 | 521 | |||
BC001797.1 | - | 384 | ||||
BC010011.2 | - | 3 | 361 | 4309613 |