Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LARP4 SIRNA Assays ›
Gene Symbol:
LARP4
Gene Name:
La ribonucleoprotein domain family member 4
Gene Aliases:
PP13296
Chromosome Location:
Chr.12: 50400809 - 50480005 on Build GRCh38
UniGene ID:
Hs.26613
Species:
Human
siRNA ID
HSS132741
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LARP4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170803.1
NP_001164274.1
14
1904
NM_001170808.1
NP_001164279.1
15
2117
NM_052879.4
NP_443111.4
16
2330
NM_199188.2
NP_954658.2
16
2327
NM_199190.2
NP_954660.1
15
2117
XM_005268613.2
XP_005268670.1
17
2379
XM_011537828.2
XP_011536130.1
17
2434
XM_011537829.2
XP_011536131.1
17
2415
XM_011537830.2
XP_011536132.1
16
2262
XM_011537831.2
XP_011536133.1
16
2326
XM_011537833.2
XP_011536135.1
16
2221
XM_011537834.2
XP_011536136.1
16
2222
XM_011537835.2
XP_011536137.1
16
2202
XM_011537836.2
XP_011536138.1
16
2203
XM_011537838.2
XP_011536140.1
16
2174
XM_011537839.2
XP_011536141.1
17
2318
XM_011537840.2
XP_011536142.1
17
2371
XM_011537841.2
XP_011536143.1
16
2224
XM_011537843.2
XP_011536145.1
15
1990
XM_011537844.2
XP_011536146.1
16
2313
XM_017018750.1
XP_016874239.1
15
2043
XM_017018751.1
XP_016874240.1
17
2195
XM_017018752.1
XP_016874241.1
16
2178
XM_017018753.1
XP_016874242.1
16
2211
XM_017018754.1
XP_016874243.1
17
2361
XM_017018755.1
XP_016874244.1
15
2028
XM_017018756.1
XP_016874245.1
16
2080
XM_017018757.1
XP_016874246.1
17
2250
XM_017018758.1
XP_016874247.1
16
2186
XM_017018759.1
XP_016874248.1
15
2011
XM_017018760.1
XP_016874249.1
15
2208
XM_017018761.1
XP_016874250.1
14
1988
XM_017018762.1
XP_016874251.1
14
1988
GenBank mRNA
AF370416.1
-
16
2208
AK055521.1
-
14
1818
AK125113.1
-
15
2120
AK291746.1
-
16
2199
AL832198.1
-
15
2058
AL833272.1
-
16
2240
BC017395.1
-
2
358
BC022377.1
-
14
1804
BC032444.2
-
3
585
BX647457.1
-
15
1941
CR936626.1
-
16
2248
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
regulation of cell morphogenesis
nucleotide binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0