Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFT2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145169.2 | NP_660152.1 | 7, 8 | 564 | ||
NR_130112.1 | 8, 9 | 587 | ||||
NR_130113.1 | 6, 7 | 492 | ||||
NR_130114.1 | 6, 7 | 483 | ||||
GenBank mRNA | AF041429.1 | - | 7, 8 | 498 | ||
AK021489.1 | - | 1, 2 | 1831 | |||
AL833217.1 | - | 1, 2 | 2895 | |||
AW964442.1 | - | 6, 7 | 414 | |||
BC018969.1 | - | 7, 8 | 469 | 3959886 | ||
BC028388.1 | - | 7, 8 | 2182 | |||
BF223532.1 | - | 6, 7 | 388 | |||
BG706927.1 | - | 7, 8 | 553 | |||
BI597156.1 | - | 8, 9 | 549 |