Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052886.2 | NP_443118.1 | 2, 3 | 395 | ||
GenBank mRNA | AK172820.1 | - | 2, 3 | 311 | ||
AK312030.1 | - | 2, 3 | 302 | |||
AY007723.1 | - | 2, 3 | 372 | |||
BC012367.1 | - | 2, 3 | 395 | 3860009 | ||
CV574204.1 | - | 2, 3 | 395 |