Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003712.1 | NP_001003712.1 | 14 | 2036 | ||
NM_001319652.1 | NP_001306581.1 | 17 | 2161 | |||
NM_001319653.1 | NP_001306582.1 | 15 | 2073 | |||
NM_001319655.1 | NP_001306584.1 | 12 | 1967 | |||
NM_020841.4 | NP_065892.1 | 15 | 2082 | |||
XM_005268621.4 | XP_005268678.1 | 18 | 2180 | |||
XM_006719224.2 | XP_006719287.1 | 14 | 2041 | |||
XM_011537855.2 | XP_011536157.1 | 15 | 1699 | |||
XM_011537856.2 | XP_011536158.1 | 16 | 2117 | |||
XM_011537857.2 | XP_011536159.1 | 18 | 2171 | |||
XM_017018768.1 | XP_016874257.1 | 15 | 1714 | |||
XM_017018769.1 | XP_016874258.1 | 16 | 2085 | |||
GenBank mRNA | AB040884.3 | - | 15 | 1753 | ||
AF323730.1 | - | 14 | 2036 | |||
AF392452.1 | - | 15 | 2083 | |||
AK092160.1 | - | 12 | 1968 | |||
AK289997.1 | - | 15 | 1802 | |||
AL832876.1 | - | 2 | 138 | |||
BC012935.1 | - | 1 | 67 | |||
BC082235.1 | - | 15 | 2021 | |||
BC093834.1 | - | 14 | 1681 | 7939679 | ||
BC101529.1 | - | 14 | 1681 | 8069035 | ||
BC111728.1 | - | 15 | 1976 | 40029118 | ||
BX647211.1 | - | 14 | 1638 | |||
CR749542.1 | - | 17 | 1886 |