Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OSBPL9 SIRNA Assays ›
Gene Symbol:
OSBPL9
Gene Name:
oxysterol binding protein like 9
Gene Aliases:
ORP-9, ORP9
Chromosome Location:
Chr.1: 51616874 - 51789219 on Build GRCh38
UniGene ID:
Hs.21938
Species:
Human
siRNA ID
HSS132914
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OSBPL9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024586.5
NP_078862.4
23
2253
NM_148904.3
NP_683702.1
19
1970
NM_148905.3
NP_683703.1
18
1874
NM_148906.2
NP_683704.2
22
2202
NM_148907.2
NP_683705.1
19
1847
NM_148908.3
NP_683706.3
22
2214
NM_148909.3
NP_683707.3
23
2283
NR_036662.1
22
2174
XM_006710318.2
XP_006710381.1
22
2013
XM_006710320.3
XP_006710383.1
20
1850
XM_006710321.2
XP_006710384.1
19
1846
XM_006710322.3
XP_006710385.1
20
2007
XM_006710323.2
XP_006710386.1
19
1915
XM_006710324.3
XP_006710387.1
21
1925
XM_006710325.3
XP_006710388.1
20
1895
XM_006710326.2
XP_006710389.1
18
1897
XM_011540599.2
XP_011538901.1
24
2107
XM_011540600.2
XP_011538902.1
23
2074
XM_011540601.2
XP_011538903.1
22
2035
XM_011540602.2
XP_011538904.1
23
2009
XM_011540603.2
XP_011538905.1
23
2033
XM_011540604.2
XP_011538906.1
20
1808
XM_017000218.1
XP_016855707.1
21
1974
XM_017000219.1
XP_016855708.1
22
1994
XM_017000220.1
XP_016855709.1
19
1735
XM_017000221.1
XP_016855710.1
19
1686
XM_017000222.1
XP_016855711.1
23
2219
XM_017000223.1
XP_016855712.1
23
2217
XM_017000224.1
XP_016855713.1
20
2162
XM_017000225.1
XP_016855714.1
21
2013
GenBank mRNA
AB208851.1
-
7
3650
AF392445.1
-
23
2027
AK022554.1
-
18
1654
AK027535.1
-
22
1984
AK027707.1
-
19
1810
AK056617.1
-
23
2072
AK074392.1
-
9
786
AK091703.1
-
22
1956
AK124037.1
-
14
2500
AK128043.1
-
25
2477
AK297656.1
-
20
2000
AK315167.1
-
19
1843
AY178997.1
-
22
1976
BC006160.2
-
3
186
BC025978.2
-
19
1880
3621193
CR457288.1
-
16
1521
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
lipid transport
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-lzswf:80/10.239.160.35:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0