Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OSBPL9 SIRNA Assays ›
Gene Symbol:
OSBPL9
Gene Name:
oxysterol binding protein like 9
Gene Aliases:
ORP-9, ORP9
Chromosome Location:
Chr.1: 51616874 - 51789219 on Build GRCh38
UniGene ID:
Hs.21938
Species:
Human
siRNA ID
HSS132915
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OSBPL9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024586.5
NP_078862.4
24
2409
NM_148904.3
NP_683702.1
20
2126
NM_148905.3
NP_683703.1
19
2030
NM_148906.2
NP_683704.2
23
2358
NM_148907.2
NP_683705.1
20
2003
NM_148908.3
NP_683706.3
23
2370
NM_148909.3
NP_683707.3
24
2439
NR_036662.1
23
2330
XM_006710318.2
XP_006710381.1
23
2169
XM_006710320.3
XP_006710383.1
21
2006
XM_006710321.2
XP_006710384.1
20
2002
XM_006710322.3
XP_006710385.1
21
2163
XM_006710323.2
XP_006710386.1
20
2071
XM_006710324.3
XP_006710387.1
22
2081
XM_006710325.3
XP_006710388.1
21
2051
XM_006710326.2
XP_006710389.1
19
2053
XM_011540599.2
XP_011538901.1
25
2263
XM_011540600.2
XP_011538902.1
24
2230
XM_011540601.2
XP_011538903.1
23
2191
XM_011540602.2
XP_011538904.1
24
2165
XM_011540603.2
XP_011538905.1
24
2189
XM_011540604.2
XP_011538906.1
21
1964
XM_017000218.1
XP_016855707.1
22
2130
XM_017000219.1
XP_016855708.1
23
2150
XM_017000220.1
XP_016855709.1
20
1891
XM_017000221.1
XP_016855710.1
20
1842
XM_017000222.1
XP_016855711.1
24
2375
XM_017000223.1
XP_016855712.1
24
2373
XM_017000224.1
XP_016855713.1
21
2318
XM_017000225.1
XP_016855714.1
22
2169
GenBank mRNA
AB208851.1
-
8
3806
AF392445.1
-
24
2183
AK022554.1
-
19
1810
AK027535.1
-
23
2140
AK027707.1
-
20
1966
AK056617.1
-
24
2228
AK074392.1
-
10
942
AK091703.1
-
23
2112
AK124037.1
-
15
2656
AK128043.1
-
26
2633
AK297656.1
-
21
2156
AK315167.1
-
20
1999
AY178997.1
-
23
2132
BC006160.2
-
4
342
BC025978.2
-
20
2036
3621193
CR457288.1
-
17
1677
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
lipid transport
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline