Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174060.1 | NP_001167531.1 | 7 | 1834 | ||
NM_017784.4 | NP_060254.2 | 8 | 2026 | |||
XM_005264843.3 | XP_005264900.1 | 8 | 1802 | |||
XM_005264844.4 | XP_005264901.1 | 7 | 1610 | |||
XM_005264845.1 | XP_005264902.1 | 3 | 710 | |||
XM_011533326.2 | XP_011531628.1 | 8 | 1179 | |||
XM_017005667.1 | XP_016861156.1 | 8 | 1692 | |||
XM_017005668.1 | XP_016861157.1 | 8 | 1669 | |||
XM_017005669.1 | XP_016861158.1 | 8 | 1198 | |||
XM_017005670.1 | XP_016861159.1 | 5 | 1881 | |||
GenBank mRNA | AB209872.1 | - | 5 | 661 | ||
AF392451.1 | - | 9 | 1736 | |||
AK000370.1 | - | 6 | 902 | |||
AK304067.1 | - | 7 | 1287 | |||
AL833247.1 | - | 8 | 1129 | |||
BC003168.1 | - | 8 | 1430 | 2905981 |