Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037131.2 | NP_001032208.1 | 7, 8 | 1393 | ||
NM_001244888.1 | NP_001231817.1 | 7, 8 | 1393 | |||
NM_014914.4 | NP_055729.2 | 7, 8 | 1393 | |||
XM_005246059.4 | XP_005246116.1 | 8, 9 | 858 | |||
XM_006712234.3 | XP_006712297.1 | 8, 9 | 859 | |||
XM_006712235.3 | XP_006712298.1 | 7, 8 | 983 | |||
XM_006712237.3 | XP_006712300.2 | 7, 8 | 1640 | |||
XM_006712239.3 | XP_006712302.2 | 8, 9 | 1655 | |||
XM_011510547.2 | XP_011508849.1 | 8, 9 | 1651 | |||
XM_011510548.2 | XP_011508850.1 | 8, 9 | 1653 | |||
XM_011510549.2 | XP_011508851.1 | 8, 9 | 1691 | |||
XM_017003282.1 | XP_016858771.1 | 7, 8 | 1529 | |||
GenBank mRNA | AB029022.1 | - | 7, 8 | 1366 | ||
AF413078.1 | - | 7, 8 | 795 | |||
AK300763.1 | - | 3, 4 | 333 | |||
AY033765.1 | - | 7, 8 | 1356 | |||
BC060814.1 | - | 7, 8 | 1393 | |||
BC140856.1 | - | 8, 9 | 1010 | 9021724 | ||
BC167153.1 | - | 7, 8 | 1366 | 8862550 | ||
BC167157.1 | - | 7, 8 | 1378 | 8862571 | ||
KJ534764.1 | - | 8, 9 | 807 |