Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRGPRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303615.1 | NP_001290544.1 | 3 | 992 | ||
NM_054030.3 | NP_473371.1 | 2 | 682 | |||
GenBank mRNA | BC063450.1 | - | 2 | 682 | 5170163 |
Set Membership: |
![]() |