Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203298.2 | NP_976043.1 | 3 | 319 | ||
GenBank mRNA | AK098720.1 | - | 2 | 956 | ||
BC015387.1 | - | 3 | 299 | 4400374 | ||
BC020852.1 | - | 3 | 306 | 4242977 | ||
BM787401.1 | - | 3 | 320 |