Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC36A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286139.1 | NP_001273068.1 | 11 | 1401 | ||
NM_152313.3 | NP_689526.2 | 11 | 1364 | |||
XM_017017174.1 | XP_016872663.1 | 11 | 1355 | |||
XM_017017175.1 | XP_016872664.1 | 10 | 1281 | |||
XM_017017176.1 | XP_016872665.1 | 10 | 1233 | |||
XM_017017177.1 | XP_016872666.1 | 10 | 1124 | |||
GenBank mRNA | AJ295983.1 | - | 6 | 750 | ||
AK096251.1 | - | 11 | 1400 | |||
AY162216.1 | - | 11 | 1318 | |||
BC047374.1 | - | 11 | 1331 | 5313230 |