Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144594.2 | NP_653195.2 | 4 | 264 | ||
XM_017018800.1 | XP_016874289.1 | 4 | 237 | |||
GenBank mRNA | AK057504.1 | - | 4 | 1074 | ||
AK098819.1 | - | 4 | 267 | |||
BC021179.1 | - | 4 | 230 | 4823487 | ||
BC105587.1 | - | 4 | 265 | 4697882 |