Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135175.1 | NP_001128647.1 | 11 | 1521 | ||
NM_001135176.1 | NP_001128648.1 | 11 | 1440 | |||
NM_001135177.1 | NP_001128649.1 | 10 | 1454 | |||
NM_152905.3 | NP_690869.1 | 12 | 1693 | |||
XM_005268644.2 | XP_005268701.1 | 12 | 1427 | |||
XM_006719237.3 | XP_006719300.1 | 10 | 1183 | |||
XM_011537903.2 | XP_011536205.1 | 9 | 1191 | |||
XM_011537904.2 | XP_011536206.1 | 12 | 1516 | |||
XM_011537905.2 | XP_011536207.1 | 11 | 1322 | |||
XM_011537906.2 | XP_011536208.1 | 11 | 1317 | |||
XM_017018801.1 | XP_016874290.1 | 11 | 1565 | |||
XM_017018802.1 | XP_016874291.1 | 11 | 1560 | |||
GenBank mRNA | AK093221.1 | - | 12 | 1631 | ||
AK290058.1 | - | 12 | 1693 | |||
AK303656.1 | - | 10 | 1420 | |||
AK315821.1 | - | 12 | 1659 | |||
AL832466.1 | - | 11 | 1407 | |||
AL832468.1 | - | 9 | 1366 | |||
BC020799.1 | - | 12 | 1653 | |||
BC027605.2 | - | 11 | 1521 | 4827191 |