Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AEBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114176.1 | NP_001107648.1 | 1, 2 | 699 | ||
NM_153207.4 | NP_694939.2 | 1, 2 | 699 | |||
XM_006719031.3 | XP_006719094.1 | 1, 2 | 881 | |||
XM_011520560.2 | XP_011518862.1 | 1, 2 | 881 | |||
GenBank mRNA | AB209384.1 | - | 1, 2 | 493 | ||
AK126973.1 | - | 2, 3 | 485 | |||
AK131361.1 | - | 1, 2 | 699 | |||
BC015624.2 | - | 1, 2 | 186 | 4843866 | ||
BC022220.1 | - | 1, 2 | 674 | 3914323 |