Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139015.4 | NP_620584.2 | 8 | 1117 | ||
XM_011537925.2 | XP_011536227.1 | 8 | 647 | |||
GenBank mRNA | AB252457.1 | - | 8 | 709 | ||
AJ345030.1 | - | 8 | 685 | |||
AJ420898.1 | - | 8 | 656 | |||
AK074916.1 | - | 7 | 600 | |||
AK223590.1 | - | 8 | 1113 | |||
AK291161.1 | - | 8 | 939 | |||
AY169313.1 | - | 8 | 891 | |||
BC025781.1 | - | 5 | 392 | |||
BC073910.1 | - | 8 | 898 | 4538522 | ||
BC101625.1 | - | 8 | 712 | 8069131 | ||
BC101627.1 | - | 8 | 712 | 8069133 |