Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173846.4 | NP_776245.2 | 2 | 262 | ||
XM_005267324.4 | XP_005267381.1 | 2 | 323 | |||
XM_011536416.1 | XP_011534718.1 | 2 | 198 | |||
XM_011536417.2 | XP_011534719.1 | 2 | 198 | |||
XM_011536418.1 | XP_011534720.1 | 2 | 198 | |||
XM_011536420.2 | XP_011534722.1 | 2 | 198 | |||
XM_017020966.1 | XP_016876455.1 | 2 | 226 | |||
GenBank mRNA | AK093070.1 | - | 2 | 194 | ||
AK098680.1 | - | 2 | 191 | |||
AY072034.1 | - | 2 | 199 | |||
AY173946.1 | - | 2 | 199 | |||
BC027610.1 | - | 2 | 267 | |||
BC038970.2 | - | 1 | 151 | 5169309 | ||
DB458250.1 | - | 2 | 262 |