Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206670.1 | NP_001193599.1 | 16 | 1815 | ||
XM_011521237.2 | XP_011519539.1 | 16 | 1725 | |||
XM_011521238.1 | XP_011519540.1 | 7 | 1533 | |||
XM_011521239.1 | XP_011519541.1 | 5 | 661 | |||
GenBank mRNA | AK127558.1 | - | 6 | 1104 | ||
BC101584.1 | - | 5 | 720 | |||
BC101612.1 | - | 5 | 720 | |||
BX648482.1 | - | 6 | 1014 |