Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144601.3 | NP_653202.1 | 4, 5 | 714 | ||
NM_181553.2 | NP_853531.1 | 3, 4 | 550 | |||
NR_037613.1 | 4, 5 | 934 | ||||
XM_006721131.1 | XP_006721194.1 | 4, 5 | 758 | |||
GenBank mRNA | AF479813.1 | - | 4, 5 | 688 | ||
AK056324.1 | - | 4, 5 | 934 | |||
AK075075.1 | - | 2, 3 | 247 | |||
BC023509.1 | - | 3, 4 | 456 | 3960441 | ||
BC023591.2 | - | 3, 4 | 448 | 4638047 | ||
BC036042.1 | - | 3, 4 | 510 | 5271808 | ||
BC040227.1 | - | 3, 4 | 501 | 5187826 | ||
BC047701.1 | - | 3, 4 | 550 | 5770132 | ||
BI457147.1 | - | 3, 4 | 402 |