Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: WDR63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288563.1 | NP_001275492.1 | 19 | 2296 | ||
NM_145172.4 | NP_660155.2 | 20 | 2413 | |||
GenBank mRNA | AK054629.1 | - | 19 | 2250 | ||
AK292603.1 | - | 20 | 2384 | |||
AY049724.1 | - | 20 | 2394 | |||
BC040265.1 | - | 20 | 2367 | 5165779 | ||
BX648851.1 | - | 20 | 2419 |