Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AXDND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144696.5 | NP_653297.3 | 17 | 2263 | ||
NR_073544.1 | 16 | 1902 | ||||
XM_011509165.1 | XP_011507467.1 | 17 | 2242 | |||
XM_011509166.2 | XP_011507468.1 | 17 | 2289 | |||
XM_011509167.2 | XP_011507469.1 | 17 | 2404 | |||
XM_011509168.1 | XP_011507470.1 | 17 | 2246 | |||
XM_011509169.1 | XP_011507471.1 | 16 | 2035 | |||
XM_011509170.1 | XP_011507472.1 | 17 | 2242 | |||
XM_011509171.1 | XP_011507473.1 | 16 | 2060 | |||
XM_011509174.1 | XP_011507476.1 | 17 | 2242 | |||
XM_011509175.1 | XP_011507477.1 | 17 | 2242 | |||
XM_011509176.1 | XP_011507478.1 | 17 | 2242 | |||
XM_011509178.2 | XP_011507480.1 | 17 | 2242 | |||
XM_011509179.2 | XP_011507481.1 | 12 | 1350 | |||
XM_011509180.1 | XP_011507482.1 | 17 | 2242 | |||
XM_011509181.2 | XP_011507483.1 | 7 | 859 | |||
XM_017000257.1 | XP_016855746.1 | 11 | 1210 | |||
XM_017000258.1 | XP_016855747.1 | 11 | 1242 | |||
XM_017000259.1 | XP_016855748.1 | 3 | 412 | |||
GenBank mRNA | AK057502.1 | - | 16 | 1773 | ||
AK302972.1 | - | 18 | 2385 | |||
AL833914.1 | - | 15 | 1690 | |||
BC146823.1 | - | 16 | 1903 | 8860438 | ||
BX647935.1 | - | 17 | 2264 |