Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRIQ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105659.1 | NP_001099129.1 | 3 | 566 | ||
NM_001322315.1 | NP_001309244.1 | 3 | 566 | |||
XM_011540663.2 | XP_011538965.1 | 3 | 580 | |||
XM_017000286.1 | XP_016855775.1 | 3 | 580 | |||
XM_017000287.1 | XP_016855776.1 | 3 | 580 | |||
XM_017000288.1 | XP_016855777.1 | 3 | 580 | |||
GenBank mRNA | AK131476.2 | - | 3 | 523 | ||
BC020778.1 | - | 3 | 526 | |||
BX647210.1 | - | 2 | 1100 |