Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138283.1 | NP_612140.1 | 2 | 423 | ||
GenBank mRNA | AY707749.1 | - | 2 | 423 | ||
BC117396.1 | - | 1 | 223 | 40125947 | ||
BI560738.1 | - | 2 | 429 | |||
FJ895879.1 | - | 1 | 48 | |||
FJ895880.1 | - | 1 | 48 |