Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042439.2 | NP_001035904.1 | 3 | 603 | ||
NM_178466.4 | NP_848561.2 | 4 | 711 | |||
GenBank mRNA | AI638623.1 | - | 4 | 519 | ||
BC066354.1 | - | 3 | 561 | 4826101 |