Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144629.2 | NP_653230.2 | 8 | 1609 | ||
XM_011510595.2 | XP_011508897.1 | 8 | 1799 | |||
XM_011510596.2 | XP_011508898.1 | 8 | 1799 | |||
XM_011510597.2 | XP_011508899.1 | 8 | 1799 | |||
XM_011510598.2 | XP_011508900.1 | 8 | 1748 | |||
XM_017003332.1 | XP_016858821.1 | 8 | 1748 | |||
GenBank mRNA | AK055136.1 | - | 8 | 1609 | ||
BC093966.1 | - | 8 | 1256 | 7939811 | ||
BC111069.1 | - | 8 | 1741 | 5764096 | ||
BC113361.1 | - | 8 | 1256 | 8322413 | ||
BX647618.1 | - | 5 | 636 |