Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301684.1 | NP_001288613.1 | 11 | 1621 | ||
NM_138803.3 | NP_620158.3 | 13 | 1877 | |||
XM_005246319.3 | XP_005246376.1 | 14 | 1764 | |||
XM_011510638.2 | XP_011508940.1 | 15 | 1804 | |||
XM_017003392.1 | XP_016858881.1 | 14 | 1916 | |||
XM_017003393.1 | XP_016858882.1 | 14 | 1599 | |||
XM_017003394.1 | XP_016858883.1 | 13 | 1749 | |||
XM_017003395.1 | XP_016858884.1 | 13 | 1812 | |||
XM_017003396.1 | XP_016858885.1 | 12 | 1773 | |||
XM_017003397.1 | XP_016858886.1 | 10 | 1250 | |||
XM_017003398.1 | XP_016858887.1 | 8 | 1245 | |||
GenBank mRNA | AF458588.1 | - | 13 | 1826 | ||
AK123220.1 | - | 8 | 1106 | |||
BC105928.1 | - | 12 | 1590 | |||
BC107596.1 | - | 11 | 1434 | 40025915 | ||
BC107597.1 | - | 13 | 1689 | 40025920 |