Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144638.2 | NP_653239.1 | 3 | 376 | ||
GenBank mRNA | AL532443.3 | - | 4 | 367 | ||
AL834253.1 | - | 2 | 2461 | |||
BC019851.1 | - | 3 | 369 | 5114515 | ||
DB499198.1 | - | 3 | 376 |