Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144643.3 | NP_653244.2 | 21 | 2549 | ||
XM_017007716.1 | XP_016863205.1 | 21 | 2561 | |||
XM_017007719.1 | XP_016863208.1 | 21 | 2082 | |||
XM_017007720.1 | XP_016863209.1 | 22 | 2461 | |||
XM_017007721.1 | XP_016863210.1 | 15 | 1663 | |||
GenBank mRNA | AK055217.1 | - | 21 | 2549 | ||
AK093361.1 | - | 15 | 1702 | |||
AK122852.1 | - | 11 | 3174 | |||
BC121057.1 | - | 12 | 1212 | |||
BC121058.1 | - | 8 | 813 | |||
BC128051.1 | - | 12 | 1211 | |||
CD679143.1 | - | 4 | 314 |