Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115007.2 | NP_001108479.1 | 5 | 364 | ||
NM_001115008.2 | NP_001108480.1 | 5 | 565 | |||
NM_001288997.1 | NP_001275926.1 | 5 | 302 | |||
NM_194282.3 | NP_919258.2 | 5 | 1228 | |||
NR_110253.1 | 3 | 1104 | ||||
NR_110254.1 | 4 | 561 | ||||
XM_005262750.4 | XP_005262807.1 | 4 | 1037 | |||
XM_006714081.3 | XP_006714144.1 | 4 | 963 | |||
XM_017007729.1 | XP_016863218.1 | 4 | 1170 | |||
XM_017007730.1 | XP_016863219.1 | 4 | 559 | |||
GenBank mRNA | AB111889.1 | - | 4 | 548 | ||
AK292769.1 | - | 5 | 312 | |||
AK294256.1 | - | 5 | 1157 | |||
BC109277.2 | - | 3 | 980 | |||
BX537919.1 | - | 4 | 462 | |||
BX640657.1 | - | 5 | 291 | |||
BX640966.1 | - | 5 | 1223 |