Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PACRGL SIRNA Assays ›
Gene Symbol:
PACRGL
Gene Name:
PARK2 coregulated like
Gene Aliases:
C4orf28
Chromosome Location:
Chr.4: 20696282 - 20757766 on Build GRCh38
UniGene ID:
Hs.479298
Species:
Human
siRNA ID
HSS134453
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PACRGL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130727.1
NP_001124199.1
3
469
NM_001258345.1
NP_001245274.1
3
469
NM_001258346.1
NP_001245275.1
3
260
NM_001317849.1
NP_001304778.1
3
429
NM_145048.3
NP_659485.1
3
469
NR_047661.1
3
293
NR_133933.1
3
326
XM_011513780.1
XP_011512082.1
5
632
XM_011513781.1
XP_011512083.1
5
451
XM_011513784.1
XP_011512086.1
5
468
XM_011513785.1
XP_011512087.1
4
551
XM_011513786.2
XP_011512088.1
5
632
XM_011513787.1
XP_011512089.1
3
423
XM_011513788.1
XP_011512090.1
4
540
XM_011513791.1
XP_011512093.1
3
383
XM_011513792.1
XP_011512094.1
3
378
XM_011513794.1
XP_011512096.1
4
331
XM_011513795.1
XP_011512097.1
3
253
XM_011513796.1
XP_011512098.1
3
248
XM_011513797.2
XP_011512099.1
3
254
XM_011513798.2
XP_011512100.1
3
190
XM_011513804.1
XP_011512106.1
3
215
XM_011513805.2
XP_011512107.1
3
254
XM_017007747.1
XP_016863236.1
5
468
XM_017007748.1
XP_016863237.1
4
338
XM_017007749.1
XP_016863238.1
3
253
XM_017007750.1
XP_016863239.1
4
540
XM_017007751.1
XP_016863240.1
4
536
XM_017007752.1
XP_016863241.1
3
383
XM_017007753.1
XP_016863242.1
3
253
XM_017007754.1
XP_016863243.1
3
469
XM_017007755.1
XP_016863244.1
3
286
XM_017007756.1
XP_016863245.1
3
279
XM_017007757.1
XP_016863246.1
3
215
XM_017007758.1
XP_016863247.1
3
299
XM_017007759.1
XP_016863248.1
3
247
GenBank mRNA
AK098692.1
-
3
326
AK291544.1
-
3
425
AK294077.1
-
3
215
AK297556.1
-
3
280
AK307092.1
-
3
240
AK307355.1
-
3
248
AK307786.1
-
5
468
AK315482.1
-
3
424
BC023002.2
-
3
216
4992139
BM763154.1
-
3
424
BX344401.2
-
3
364
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0