Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SVOPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174959.3 | NP_777619.1 | 1, 2 | 90 | ||
XM_011515797.2 | XP_011514099.1 | 2, 3 | 164 | |||
XM_017011747.1 | XP_016867236.1 | 2, 3 | 104 | |||
XM_017011748.1 | XP_016867237.1 | 2, 3 | 109 | |||
GenBank mRNA | BC036796.1 | - | 1, 2 | 90 | 5590416 |