Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178819.3 | NP_848934.1 | 4 | 1229 | ||
XM_011544392.2 | XP_011542694.1 | 5 | 1352 | |||
XM_011544393.1 | XP_011542695.1 | 4 | 1241 | |||
XM_011544394.1 | XP_011542696.1 | 4 | 1278 | |||
XM_017013042.1 | XP_016868531.1 | 5 | 1379 | |||
XM_017013043.1 | XP_016868532.1 | 4 | 1227 | |||
GenBank mRNA | AF317516.1 | - | 3 | 455 | ||
AF406612.1 | - | 4 | 1217 | |||
AY358670.1 | - | 3 | 366 | |||
AY513610.1 | - | 3 | 449 | |||
BC051377.1 | - | 3 | 409 | 6649895 | ||
BC061884.1 | - | 3 | 409 | 6577569 |