Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBLN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080617.5 | NP_542184.1 | 3 | 1767 | ||
GenBank mRNA | AK289640.1 | - | 3 | 1571 | ||
AY358527.1 | - | 3 | 802 | |||
BC050026.1 | - | 3 | 1769 | 4820943 | ||
BC069402.1 | - | 3 | 466 | 7262170 | ||
BC069488.1 | - | 3 | 466 | 7262182 | ||
BC069516.1 | - | 3 | 466 | 7262099 |