Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301144.1 | NP_001288073.1 | 2 | 419 | ||
NM_001301145.1 | NP_001288074.1 | 2 | 419 | |||
NM_001301146.1 | NP_001288075.1 | 2 | 419 | |||
NM_080745.4 | NP_542783.2 | 2 | 419 | |||
NM_182985.4 | NP_892030.3 | 3 | 896 | |||
GenBank mRNA | AF302088.1 | - | 2 | 267 | ||
AF302089.1 | - | 2 | 267 | |||
AK226115.1 | - | 2 | 2528 | |||
AK292252.1 | - | 3 | 740 | |||
AL360161.1 | - | 2 | 254 | |||
AY305385.1 | - | 3 | 594 | |||
BC024199.1 | - | 3 | 714 | |||
BC033314.1 | - | 2 | 215 | 5166812 | ||
BC047945.1 | - | 3 | 733 | 5744224 | ||
BI829233.1 | - | 3 | 712 | |||
BM559467.1 | - | 3 | 742 |