Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301144.1 | NP_001288073.1 | 3 | 633 | ||
NM_080745.4 | NP_542783.2 | 3 | 625 | |||
NM_182985.4 | NP_892030.3 | 4 | 1102 | |||
GenBank mRNA | AF302088.1 | - | 3 | 481 | ||
AK226115.1 | - | 2 | 4889 | |||
AK292252.1 | - | 4 | 946 | |||
AL360161.1 | - | 3 | 468 | |||
AY305385.1 | - | 4 | 800 | |||
BC024199.1 | - | 4 | 920 | |||
BC033314.1 | - | 3 | 421 | 5166812 | ||
BC047945.1 | - | 4 | 939 | 5744224 |