Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMCR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144775.2 | NP_658988.2 | 1 | 1911 | ||
GenBank mRNA | AF467440.1 | - | 1 | 1789 | ||
AK296847.1 | - | 1 | 1899 | |||
BC101115.1 | - | 1 | 1785 | |||
BC101116.2 | - | 1 | 1785 | |||
BC101117.2 | - | 1 | 1785 | |||
BC142680.1 | - | 1 | 1881 | 40148405 |